Order Entry
Italy
ContactUsLinkComponent
Human Recombinant tPA (From CHO cells)
Human Recombinant tPA (From CHO cells)
Catalog # USBIT5600-07B
Supplier:  US Biological
CAS Number:  
Human Recombinant tPA (From CHO cells)
Catalog # USBIT5600-07B
Supplier:  US Biological
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage conditions:
    Aliquot to avoid repeated freezing and thawing and store at −70 °C. Aliquots are stable for 6 months after receipt. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.
  • Form:
    Supplied as a liquid in 0,5 M HEPES, 0,5 M sodium chloride
  • Protein/peptide name:
    tPA (Tissue Plasminogen Activator)
  • Purity:
    ≥95% (SDS-PAGE)
  • Solubility:
    >2 mg/ml
  • Molecular weight:
    ~70 kD
  • pH:
    7,4
  • Shipping temperature:
    Dry Ice

Specifications

About this item

This recombinant human tPA is active and expressed in Chinese hamster ovary (Cho) cells. Fully complexable with PAI-1.

  • Activity: >90% active as determined by titration with PAI-1
Caution: Intended for research use only. Not for use in human, therapeutic, or diagnostic applications.