Order Entry
Italy
ContactUsLinkComponent
Iodine 0.02 M in THF : pyridine : water (69:22:11) (w:w:w), Novabiochem® for DNA/RNA synthesis, oxidizing reagent B
Flammable
Health
Irritant
Iodine 0.02 M in THF : pyridine : water (69:22:11) (w:w:w), Novabiochem® for DNA/RNA synthesis, oxidizing reagent B
Catalog # NOVA8.57013.4000
CAS Number:  
Flammable
Health
Irritant
Iodine 0.02 M in THF : pyridine : water (69:22:11) (w:w:w), Novabiochem® for DNA/RNA synthesis, oxidizing reagent B
Catalog # NOVA8.57013.4000
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

About this item

 :
 :253.81 g/mol
ADR:3,II
 :M-Clarity™ quality level = MQ200
Flash Pt:–21.5 °C
UN:1993
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355