Pharmaceutical Standards, Dihydro Cyclosporin A
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
Specifications
- CAS No.:59865-15-5
- Shipping conditions:RT
- Molecular weight:1204,63
- Storage temperature:+4 °C
Specifications
Frequently Bought Together