
[(1,2,5,6-η)-1,5-Cyclooctadiene][(2,3,5,6-η)-2,3,5,6-tetramethyl-2,5-cyclohexadiene-1,4-dione]nickel ≥98.0% (by titration analysis on dried basis)
: TCI
:40759-64-6
:
:MFCD32851669
:227 °C (dec.)
:GHS08
:Room Temperature
- Appearance:Red to Dark red powder to crystal
- Purity(Chelometric Titration):min. 98.0 %
- NMR:confirm to structure
Frequently Bought Together