Ethanol 78% ≥78% (v/v)
VWRS83812.410
:
- Pack type:Plastic container
- Pk:10 L
- Seq:5214
- Environmentally Preferable:
- Density (20/20):0.864 0.87
- Assay:77.5 78.5 %
: Ethyl alcohol
:
:46.07 g/mol
:200-578-6
:1170
:GHS02
:Ambient
:64-17-5
:C₂H₅OH
:MFCD00003568
:78.3
:-117
:12
:3 III
:12,03806