Order Entry
Italy
Orders LinkContactUsLinkComponent
β-Alanine 99%

β-Alanine 99%

β-Alanine 99%
Certificates
β-Alanine 99%

3 Options Available Below

Product Details & Documents
Synonyms:  H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
  • CAS Number:107-95-9
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00008200
  • Molecular Weight:89.09 g/mol
  • Merck Index:13,00204
  • Boiling Pt:201 °C (1013 hPa)
  • Melting Pt:197…202 °C (dec.)
  • Density:1,44 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-536-5

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 % (105°C, 3 h)
  • Sulfated ash:
    ≤0.2 %
  • pH:
    6.5 to 7.5 (10 % w/v in water)
  • Titration with HClO4:
    ≥98.5 % (on dry substance)
  • Chloride (Cl):
    ≤400 ppm
  • Sulfate (SO4):
    ≤480 ppm
  • Ammonium (NH4):
    ≤1000 ppm
  • Iron (Fe):
    ≤30 ppm
  • Arsenic oxide (As2O3):
    ≤2 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Other amino acids:
    (TLC) Passes test (not detectable on 10 µg spotting)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 72,20 €
    72,20 €
    1 * 500 g
       72,20 €

  • Plastic bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 258,00 €
    258,00 €
    1 * 2,5 kg
       258,00 €

  • Plastic drum
    10 kg
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 766,00 €
    766,00 €
    1 * 10 kg
       766,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.