
N-(tert-Butyl)-N-fluoro-2-methylpropane-2-sulphonamide ≥95.0% (by GC), solid
: TCI
:2845105-20-4
:
:C₈H₁₈FNO₂S
:MFCD34705242
:211.30 g/mol
:33 °C
:GHS07
:Refrigerator
- Appearance:White to Light yellow powder to lump
- Purity(GC):min. 95.0 %
- NMR:confirm to structure
Frequently Bought Together