Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Furniture
Storage Cabinets
Accessories for Storage Cabinets, Tall
Accessories for Storage Cabinets, Tall
Accessories for Storage Cabinets, Tall
Catalog # KOTT961-00047
Supplier:
KOETTERMANN
Expand image
Accessories for Storage Cabinets, Tall
Catalog # KOTT961-00047
Supplier:
KOETTERMANN
Specifications
Description:
Touch-up paint colour: Arctic white (NCS S 0505 R80B) similar to RAL design 260 90 05
Pk:
1
Specifications
About this item
Product Details
Product Details