Order Entry
Italy
ContactUsLinkComponent
Anti-SARS-CoV-2 (COVID-19) Spike S1 Rabbit Polyclonal Antibody
Anti-SARS-CoV-2 (COVID-19) Spike S1 Rabbit Polyclonal Antibody
Catalog # PRSI9083
Supplier:  ProSci Inc.
undefined
Anti-SARS-CoV-2 (COVID-19) Spike S1 Rabbit Polyclonal Antibody
Catalog # PRSI9083
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SARS-CoV-2 (COVID-19) Spike S1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    9083
  • Modification:
    None
  • Storage buffer:
    SARS-CoV-2 (COVID-19) Spike S1 antibody is supplied in PBS containing 0,02% sodium azide
  • Storage temperature:
    Store at +4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles
  • Concentration:
    1 mg/ml
  • Immunogen:
    Anti-SARS-CoV-2 (COVID-19) Spike S1 antibody (9083) was raised against a peptide corresponding to 16 amino acids near the amino terminus of SARS-CoV-2 (COVID-19) Spike S1 glycoprotein
  • Purification:
    SARS-CoV-2 (COVID-19) Spike S1 antibody is affinity chromatography purified via peptide column
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: SARS-CoV-2 (COVID-19) Spike S1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus