Crystallising Dishes, without Spout
Supplier: TECHNICAL QUARTZ SOLUTIONS
Certificates
About this item
Temperature-resistant crystallising dishes made from transparent quartz glass.
- Excellent chemical resistance
- High resistance against thermal shock
- Made for tasks up to 1050 °C
- High purity
The TQS quartz glass crystallising dishes are made for tasks up to 1050 °C (maximum limit 1280 °C). High chemical resistance and purity characterise these products. Quartz glass products have a long life for frequent use with temperatures up to 1050 °C, but the durability is dependent upon use and applications.
Frequently Bought Together