About this item
Synonyms: (E)-1-Methoxy-4-(prop-1-en-1-yl)benzene
- CAS Number:4180-23-8
- Formula:C₁₀H₁₂O
- MDL Number:MFCD00009284
- Molecular Weight:148.2 g/mol
- Boiling Pt:236 °C (1013 hPa)
- Melting Pt:21…23 °C
- Density:0.9883 g/cm³ (20 °C)
- Flash Pt:96 °C
- Storage Temperature:Ambient
- EINECS:224-052-0
- ADR:9,III
- :M-Clarity™ quality level = MQ200
- UN:3082
2 Options Available Below
Product Details & Documents
Synonyms: (E)-1-Methoxy-4-(prop-1-en-1-yl)benzene
- CAS Number:4180-23-8
- Formula:C₁₀H₁₂O
- MDL Number:MFCD00009284
- Molecular Weight:148.2 g/mol
- Boiling Pt:236 °C (1013 hPa)
- Melting Pt:21…23 °C
- Density:0.9883 g/cm³ (20 °C)
- Flash Pt:96 °C
- Storage Temperature:Ambient
- EINECS:224-052-0
- ADR:9,III
- :M-Clarity™ quality level = MQ200
- UN:3082
Specifications for Product Family
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:21 - 23 °C
- Molar Mass:148.20 g/mol
- Boiling Point:235 - 237 °C (1013 hPa)
- Vapor Pressure:1.33 hPa (63 °C)
- Flash Point:96 °C
- Refractive Index:1.56 (20 °C, 589 nm)
- Density:0.99 g/cm3 (20 °C)
- pH:7 (H2O)



