trans-Anethole
Glass bottle for solids, 100 mg
Catalog # PROOMM0164.01
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: (E)-1-Methoxy-4-(prop-1-en-1-yl)benzene
- CAS Number:4180-23-8
- Formula:C₁₀H₁₂O
- MDL Number:MFCD00009284
- Molecular Weight:148.2 g/mol
- Boiling Pt:236 °C (1013 hPa)
- Melting Pt:21…23 °C
- Density:0,9883 g/cm³ (20 °C)
- Flash Pt:96 °C
- EINECS:224-052-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



