Order Entry
Italy
ContactUsLinkComponent
Sodium hexanitritocobaltate(III), Supelco®
Oxidizing
Health
Irritant
Sodium hexanitritocobaltate(III), Supelco®
Supplier:  Merck
Certificates
Oxidizing
Health
Irritant
Sodium hexanitritocobaltate(III), Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    720 g/l (20 °C)
  • Melting Point:
    220 °C
  • Molar Mass:
    403.93 g/mol
  • Bulk Density:
    480 kg/m3
  • Density:
    0.48 g/cm3
  • pH:
    5 (100 g/l, H2O, 20 °C)
  • Hazard Class:
    5.1
  • Hazard Class Description:
    Oxidizer

Specification Test Results

Related Information

Frequently Bought Together