Bis(2-iodophenyl)disulphide
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:87797-73-7
:
Formula:C₁₂H₈IS₂
MDL Number:MFCD00099178
Molecular Weight:343.23 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together