Order Entry
Italy
ContactUsLinkComponent
Boron trichloride 1 mol/l in dichloromethane, AcroSeal®
Toxic
Health
Corrosive
Boron trichloride 1 mol/l in dichloromethane, AcroSeal®
Certificates
undefined
Toxic
Health
Corrosive
Boron trichloride 1 mol/l in dichloromethane, AcroSeal®
Specification Test Results
  • Appearance:
    Clear colorless to brown Solution (May darken on storage)
  • Titration Argentometric:
    0.9 to 1.1 M
  • Titration:
    ≥98.5 %
  • Chloride (Cl):
    ≤0.1 %
  • Sulfate (SO4):
    ≤0.07 %
  • Insoluble matter:
    ≤0.01 %