Order Entry
Italy
ContactUsLinkComponent
Diammonium hydrogen citrate, high purity
Diammonium hydrogen citrate, high purity
Catalog # 0470-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Diammonium hydrogen citrate, high purity
Catalog # 0470-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Chloride:
    <0.003%
  • Heavy Metals:
    <0.003%
  • Purity:
    ≥99.0%
  • Residue after ignition:
    ≤0.05%
  • Sulphate:
    ≤0.01%

Specification Test Results

About this item

 :
 :226.19 g/mol
Merck Index:13,00513
Density:1.483 g/cm³ (20 °C)
EINECS:221-146-3
 :GHS07
Storage Temperature:Ambient
CAS Number:3012-65-5
Formula:HOC(CO₂H)(CH₂CO₂NH₄)₂
MDL Number:MFCD00013068