
5-Isopropyl-2-methylphenol, Sigma-Aldrich®
: Merck
:499-75-2
:
:C₁₀H₁₄O
:MFCD00002236
:237 °C (1013 hPa)
:0.5 °C
:0.98 g/cm³ (20 °C)
:>100 °C
:207-889-6
:3265
:8,III
:M-Clarity™ quality level = MQ200
:150.22 g/mol
:GHS07
:Ambient
- Solubility:1.25 g/l (25 °C)
- Melting Point:3 °C
- Molar Mass:150.22 g/mol
- Boiling Point:237 - 239 °C
- Flash Point:100 °C
- Refractive Index:1.5229 (20 °C, 589 nm)
- Density:0.98 g/cm3 (20 °C)
Frequently Bought Together