Order Entry
Italy
ContactUsLinkComponent
Pesticide reference standard, 3-Chlorotoluene 100 µg/ml in methanol, Dr Ehrenstorfer
Pesticide reference standard, 3-Chlorotoluene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA11521000ME
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 3-Chlorotoluene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA11521000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    108-41-8
  • Description:
    3-Chlorotoluene 100 µg/ml in methanol
  • Formula:
    C₇H₇Cl
  • Description:
    3-Chlorotoluene

Specifications

About this item