Order Entry
Italy
Orders LinkContactUsLinkComponent
(±)-1,2-Octanediol ≥97%

(±)-1,2-Octanediol ≥97%

(±)-1,2-Octanediol ≥97%
Certificates
(±)-1,2-Octanediol ≥97%
(±)-1,2-Octanediol ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 1117-86-8 Molecular Formula: C8H18O2 Formula Weight: 146.23 Flash Point: 138°(280°F) Melting Point: 28-34° Boiling Point: 131-133°/10mm Beilstein Reference: 1719619 MDL No.: MFCD00010738

Synonyms:  1,2-Octanediol, (±)-1,2-Octylene glycol, (±)-1,2-Dihydroxy octane
  • CAS Number:1117-86-8
  • Formula:C₈H₁₈O₂
  • MDL Number:MFCD00010738
  • Molecular Weight:146.23 g/mol
  • Boiling Pt:131…133 °C (10 mmHg)
  • Melting Pt:28…34 °C
  • Flash Pt:138 °C (280 °F)
  • Storage Temperature:Ambient
  • EINECS:214-254-7
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.