
3-Nitrophthalimide 97%, Sigma-Aldrich®
Catalog # 414034-10G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance (Colour):Conforms to Requirements
Faint to Dark Yellow - Appearance (Form):Conforms to Requirements
Powder or Crystals - Infrared spectrum:Conforms to Structure
- Carbon:48.2 - 51.8%
- Nitrogen:14.0 - 15.1%
- Purity (HPLC):≥96.5%
Specification Test Results
About this item
:
:192.13 g/mol
Melting Pt:218 °C
:M-Clarity™ quality level = MQ200
EINECS:210-051-2
:GHS07
Storage Temperature:Room Temperature
CAS Number:603-62-3
Formula:C₈H₄N₂O₄
MDL Number:MFCD00041852