Eucalyptol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:470-82-6
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00167977
:154.25 g/mol
Merck Index:12,03940
Boiling Pt:176…177 °C (1013 hPa)
Melting Pt:1.5 °C
Density:0.92 g/cm³ (20 °C)
EINECS:207-431-5
UN:1993
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:46 °C
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:3.5 g/l (21 °C)
- Melting Point:1.5 °C
- Molar Mass:154.25 g/mol
- Boiling Point:174 - 177 °C
- Flash Point:49 °C
- Density:0.925 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together