

Benzyltrimethylammonium chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:56-93-9
:
Formula:C₁₀H₁₆ClN
MDL Number:MFCD00011782
:185.7 g/mol
Melting Pt:238 °C
Density:1.07 g/cm³ (20 °C)
EINECS:200-300-3
UN:2811
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:GHS06
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:800 g/l (20 °C)
- Melting Point:243 °C
- Molar Mass:185.7 g/mol
- Bulk Density:135 °C
- Boiling Point:- 0.0001 hPa (20 °C)
Specification Test Results
Frequently Bought Together