2-Thiopheneethanol for synthesis, Sigma-Aldrich®
Catalog # 8.18682.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 ml
- Seq:2315
Specifications
Specification Test Results
- Molar Mass:128.18 g/mol
- Boiling Point:74 - 76 °C (2 hPa)
- Refractive Index:1.5478 (20 °C, 589 nm)
- Density:1.16 g/cm3 (20 °C)
- Synonyms:2-(2-Thienyl)ethanol, 2-(2-Hydroxyethyl)thiophene
- Hill Formula:C6H8OS
Specification Test Results
About this item
:
:128.19 g/mol
Boiling Pt:108 °C (1013 hPa)
Density:1.197 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:226-452-0
CAS Number:5402-55-1
Formula:C₆H₈OS
MDL Number:MFCD00005462