
2,5-Dimethyl-3-hexyne-2,5-diol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:142-30-3
:
Formula:C₈H₁₄O₂
MDL Number:MFCD00004468
:142.2 g/mol
Boiling Pt:205 °C (1013 hPa)
Melting Pt:93…95 °C
Density:0.9469 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:205-533-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:240 g/l (20 °C)
- Melting Point:93 - 95 °C
- Molar Mass:142.19 g/mol
- Boiling Point:205 °C (1013 hPa)
- Density:0.94 g/cm3 (20 °C)
- Hill Formula:C8H14O2
Specification Test Results
Frequently Bought Together