Allyl cyanoacetate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13361-32-5
:
Formula:C₆H₇NO₂
MDL Number:MFCD00013817
:125.13 g/mol
Melting Pt:–40 °C
Density:1.07 g/cm³ (20 °C)
Flash Pt:91 °C
EINECS:236-423-4
ADR:6.1,III
:M-Clarity™ quality level = MQ200
UN:3276
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:-40.0 °C
- Molar Mass:125.13 g/mol
- Boiling Point:89 - 90 °C (5 hPa)
- Vapor Pressure:0.05 hPa
- Flash Point:91 °C
- Refractive Index:1.4393 (28 °C, 589 nm)
- Density:1.072 g/cm3 (20 °C)
- Synonyms:Cyanoacetic acid allyl ester
- Hill Formula:C6H7NO2
- Chemical Formula:NCCH2COOCH2CH=CH2
Specification Test Results
Frequently Bought Together