

Valeronitrile for synthesis
8.08514.0100
: Merck
:
- Pk:100 mL
- Seq:2440
- Hill Formula:C₅H₉N
- Chemical formula:CH₃(CH₂)₃CN
- Molar Mass:83.13 g/mol
- Boiling point:141 °C (1013 mbar)
- Density:0.799 g/cm³ (20 °C)
- Flash point:38 °C
- Ignition temperature:520 °C
- Melting Point:-96 °C
- Vapor pressure:7.0 hPa (20 °C)
- Refractive Index:1.3973 (20 °C, 589 nm)
- Solubility:10.0 g/l (20 °C)
- LD 50 oral:LD50 Rat 332 mg/kg
- Storage class:3 Flammable liquids
- WGK:WGK 2 water endangering
- R Phrase:R 10-22
:
:83.13 g/mol
:12,10043
:139 °C (1013 hPa)
:–96 °C
:0.799 g/cm³ (20 °C)
:38 °C
:203-781-8
:1993
:3,III
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
:Ambient
:110-59-8
:C₅H₉N
:MFCD00001974