

2,2'-Phenyliminodiethanol for synthesis
Supplier: Merck
Certificates
About this item
CAS Number:120-07-0
:
Formula:(C₆H₅)N(CH₂CH₂OH)₂
MDL Number:MFCD00002845
:181.23 g/mol
Boiling Pt:348 °C (1013 hPa)
Melting Pt:56…59 °C
Density:1.201 g/cm³ (60 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:200 °C
Storage Temperature:Ambient
EINECS:204-368-5
:GHS05
:GHS08
Specification Test Results
- Solubility:3 g/l (20 °C)
- Melting Point:56 - 59 °C
- Molar Mass:181.23 g/mol
- Boiling Point:270 °C
- Vapor Pressure:- 1 hPa (20 °C)
- Flash Point:200 °C
- Density:1.1 g/cm3 (60 °C)
- pH:8.2 (3 g/l, H2O, 50 °C)
- Ignition Point:380 °C DIN 51794
Specification Test Results
Frequently Bought Together