Order Entry
Italy
ContactUsLinkComponent
Cinnamaldehyde for synthesis, Sigma-Aldrich®
Cinnamaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Cinnamaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    1.1 g/l (20 °C)
  • Melting Point:
    -8 °C
  • Molar Mass:
    132.16 g/mol
  • Boiling Point:
    127 °C (21 hPa)
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Flash Point:
    138 °C
  • Refractive Index:
    1.6219 (20 °C, 589 nm)
  • Density:
    1.05 g/cm3 (20 °C)
  • Synonyms:
    trans-Cinnamic aldehyde, trans-3-Phenyl-2-propenal
  • Hill Formula:
    C9H8O
  • Chemical Formula:
    C6H5CH=CHCHO

Specification Test Results

Related Information

Frequently Bought Together