
Undec-10-enoic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:112-38-9
:
Formula:C₁₁H₂₀O₂
MDL Number:MFCD00004442
:184.28 g/mol
Merck Index:12,09983
Boiling Pt:300 °C (1013 hPa)
Melting Pt:24 °C
Density:0.91 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:146 °C
EINECS:203-965-8
:GHS07
Specification Test Results
- Solubility:73.7 mg/l (30 °C)
- Melting Point:24 °C
- Molar Mass:184.27 g/mol
- Boiling Point:275 °C (1013 mbar)
- Flash Point:149 °C
- Density:0.91 g/cm3 (20 °C)
- Ignition Point:275 °C
Specification Test Results
Related Information
Frequently Bought Together