Safranine O, Certistain® for microscopy, Sigma-Aldrich®
Catalog # 1.15948.0000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Solubility:50 g/l (20 °C)
- Molar Mass:350.85 g/mol
- Bulk Density:400 kg/m3
- pH:10 (10 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:350.85 g/mol
Density:1.447 g/cm³ (22 °C)
:M-Clarity™ quality level = MQ300
EINECS:207-518-8
:GHS05
Storage Temperature:Ambient
:1
CAS Number:477-73-6
Formula:C₂₀H₁₉ClN₄
MDL Number:MFCD00011759