
Methacrylamide
Catalog # EHERC14971500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:85.11 g/mol
Boiling Pt:215 °C (1013 hPa)
Melting Pt:106…109 °C
Density:1,053 g/cm³ (20 °C)
Flash Pt:103 °C
EINECS:201-202-3
:GHS07
CAS Number:79-39-0
Formula:C₄H₇NO
MDL Number:MFCD00008018