Order Entry
Italy
ContactUsLinkComponent
Uracil, Millipore®
Uracil, Millipore®
Catalog # 6630-10
CAS Number:  
Uracil, Millipore®
Catalog # 6630-10
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :112.09 g/mol
Merck Index:12,09985
Boiling Pt:379 °C (1013 hPa)
Melting Pt:194 °C
Density:0.99744 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:200-621-9
CAS Number:66-22-8
Formula:C₄H₄N₂O₂
MDL Number:MFCD00006016