
Sodium-3-nitrobenzoate, (max. 10% H₂O) ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:827-95-2
:MFCD00051097
Formula:C₇H₄NNaO₄
MDL Number:MFCD00150535
:189.1 g/mol
Melting Pt:>300 °C
EINECS:212-578-3
:GHS06
Storage Temperature:Ambient
Related Information
Frequently Bought Together