
Benzyltripropylammonium chloride 96%
Catalog # L00675.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:269.85 g/mol
Melting Pt:∼ 190 °C (dec.)
:GHS07
Storage Temperature:Ambient
CAS Number:5197-87-5
Formula:C₁₆H₂₈ClN
MDL Number:MFCD00051869