


4-Chloro-N-methylaniline 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:932-96-7
:
Formula:C₇H₈ClN
MDL Number:MFCD00000614
:141.6 g/mol
Boiling Pt:238…240 °C
Density:1.169
Flash Pt:>100 °C (212 °F)
EINECS:213-262-8
UN:2810
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together