Order Entry
Italy
ContactUsLinkComponent
Tetramethylammonium hexafluorophosphate 99%
Toxic
Corrosive
Tetramethylammonium hexafluorophosphate 99%
Catalog # A17467.22
CAS Number:  
Toxic
Corrosive
Tetramethylammonium hexafluorophosphate 99%
Catalog # A17467.22
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

 :
 :219.11 g/mol
Melting Pt:>300 °C
Density:1.617 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:209-194-3
UN:2923
ADR:8,III
 :GHS06
 :GHS05
CAS Number:558-32-7
Formula:C₄H₁₂F₆NP
MDL Number:MFCD00012012