Eucalyptol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:470-82-6
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00167977
:154.25 g/mol
Merck Index:14,3895
Boiling Pt:176…177 °C
Melting Pt:1.0 °C
Density:0.923
Flash Pt:49 °C (120 °F)
Storage Temperature:Ambient
EINECS:207-431-5
UN:3271
ADR:3,III
:GHS02
:GHS05
:GHS07
Related Information
Frequently Bought Together