Tris(3-chlorophenyl)phosphine ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 29949-85-7 Molecular Formula: C18H12Cl3P Formula Weight: 365.63 Melting Point: 64-66° MDL No.: MFCD00013632
Synonyms: Tri(m-chlorophenyl)phosphine, Tris(m-chlorophenyl)phosphine
- CAS Number:29949-85-7
- Formula:C₁₈H₁₂Cl₃P
- MDL Number:MFCD00013632
- Molecular Weight:365.63 g/mol
- Melting Pt:64…66 °C
- Storage Temperature:Ambient
- EINECS:249-970-9
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 29949-85-7 Molecular Formula: C18H12Cl3P Formula Weight: 365.63 Melting Point: 64-66° MDL No.: MFCD00013632
Synonyms: Tri(m-chlorophenyl)phosphine, Tris(m-chlorophenyl)phosphine
- CAS Number:29949-85-7
- Formula:C₁₈H₁₂Cl₃P
- MDL Number:MFCD00013632
- Molecular Weight:365.63 g/mol
- Melting Pt:64…66 °C
- Storage Temperature:Ambient
- EINECS:249-970-9
Documents
Certificates
Search by Lot Numbermsds_ALFAA11203
Scheda di sicurezza
Organic Building Blocks and Intermediates
Note applicative
Product Family Options
Product Information
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



