

Tetrafluoroboric acid ≥48% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16872-11-0
:
Formula:HBF₄
MDL Number:MFCD00011345
:87.81 g/mol
Merck Index:14,4149
Boiling Pt:130 °C (dec.)
Density:1.4
Storage Temperature:Ambient
EINECS:240-898-3
UN:1775
ADR:8,II
:GHS06
:GHS05
Specification Test Results
- Concentration (weight/weight):48% min
Specification Test Results
Related Information
Frequently Bought Together