

Tetramethylammonium perchlorate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2537-36-2
:
Formula:C₄H₁₂ClNO₄
MDL Number:MFCD00043207
:173.6 g/mol
Melting Pt:300 °C
Density:1,379 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:219-805-5
UN:1479
ADR:5.1,II
:GHS07
:GHS03
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Assay:≥97.5 % (on dry base)
- Water:1 to 7 %
- GC:≥99.0 %
- Specific optical rotation:+39° to +41° (20°C, 589 nm) (neat)
Specification Test Results
Frequently Bought Together