

1-(4-Methylphenyl)piperazine 98%
Catalog # ACRO370830050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to yellow Low melting solid
- Infrared spectrum:Conforms
- Melting point:29 to 34 °C
- GC:≥97.5 %
- Titration with HClO4:≥97.5 %
Specification Test Results
About this item
:
:176.26 g/mol
Melting Pt:29…34 °C
EINECS:254-534-6
UN:3259
:GHS07
:GHS05
CAS Number:39593-08-3
Formula:C₁₁H₁₆N₂
MDL Number:MFCD00040737