

N,O-Bis(trimethylsilyl)trifluoroacetamide 98%
ACRO321200100
:
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3830 to 1.3850 (20°C, 589 nm)
- GC:≥97.5 %
:
:257.4 g/mol
:45…50 °C (14 mmHg)
:–10 °C
:0.96 g/cm³
:23 °C
:Refrigerator
:247-103-9
:2920
:8,II
:GHS05
:GHS02
:25561-30-2
:C₈H₁₈F₃NOSi₂
:MFCD00008269