Order Entry
Italy
ContactUsLinkComponent
(Trifluoromethyl)trimethylsilane 99%
Irritant
Flammable
(Trifluoromethyl)trimethylsilane 99%
Certificates
undefined
Irritant
Flammable
(Trifluoromethyl)trimethylsilane 99%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3290 to 1.3320 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together