
Dipotassium tetraborate tetrahydrate 99.5%, extra pure
:12045-78-2
:
:B₄K₂O₇·4H₂O
:MFCD00149924
:305.49 g/mol
:850 °C
:1,95 g/cm³ (20 °C)
:215-575-5
:GHS08
:Ambient
- Appearance:White Powder
- Titration with HClO4:≥99.4 %
- Chloride (Cl):≤20 ppm
- Sulfate (SO4):≤500 ppm
- Iron (Fe):≤20 ppm
- Heavy metals (as Pb):≤20 ppm
Frequently Bought Together