Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
N,N-Dimethylethylamine 99%
 
undefined
 
 
 
N,N-Dimethylethylamine 99%

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3715 to 1.3735 (20°C, 589 nm)
  • Titration with HClO4:
    ≥98.5 %

 

 

Frequently Bought Together