2,2,2-Trichloroethyl chloroformate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17341-93-4
:
Formula:C₃H₂Cl₄O₂
MDL Number:MFCD00000810
:211.86 g/mol
Boiling Pt:171…172 °C (760 mmHg)
Melting Pt:1 °C
Density:1.539 g/cm³
Flash Pt:>100 °C
Storage Temperature:Refrigerator
EINECS:241-363-7
UN:3277
ADR:6.1,II
:GHS06
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4680 to 1.4720 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together