Order Entry
Italy
ContactUsLinkComponent
N,N,N',N'-Tetramethyl-1,3-propanediamine 99+%
Toxic
Corrosive
Flammable
Environment
N,N,N',N'-Tetramethyl-1,3-propanediamine 99+%
Certificates
undefined
Toxic
Corrosive
Flammable
Environment
N,N,N',N'-Tetramethyl-1,3-propanediamine 99+%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4225 to 1.4245 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

Related Information

Frequently Bought Together