Order Entry
Italy
ContactUsLinkComponent
Diethylene glycol monobutyl ether 99+%
Irritant
Diethylene glycol monobutyl ether 99+%
Certificates
undefined
Irritant
Diethylene glycol monobutyl ether 99+%

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4310 to 1.4330 (20°C, 589 nm)
  • Water:
    ≤0.2 %
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Specific gravity:
    (20°C) 0.952 to 0.958
  • Acidity (CH3COOH):
    ≤0.05 %

Specification Test Results

Related Information

Frequently Bought Together