
2,3-Dimethoxyaniline ≥98.0% (by GC, titration analysis)
: TCI
:6299-67-8
:
:C₈H₁₁NO₂
:MFCD00180769
:153.18 g/mol
:93°C
:GHS07
:Refrigerator
- Appearance:Colorless to Yellow to Orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together