Order Entry
Italy
ContactUsLinkComponent
 
Irritant
Dowex® 50WX8 (H) (strongly acidic cation exchanger), Supelco®
Catalog # 1.05221.0250
Supplier:  Merck
CAS Number:  
undefined
Irritant
Dowex® 50WX8 (H) (strongly acidic cation exchanger), Supelco®
Catalog # 1.05221.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Description:
    light yellow to brown spherules
  • Particle size (0,09-0,25 mm):
    ≥ 70.0 %
  • Total capacity:
    ≥ 1.7 mol/l

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ100
 :GHS07
CAS Number:11119-67-8
 :69011-20-7
MDL Number:MFCD00132726